Human GAL1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA321HU
MSPP-SEA321HUEA 498 USD
MSPP-SEA321HU
Human GAL1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human GAL1 (Galectin 1). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.123 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Galectin-1 is one of the most important lectins to date participating in the malignant tumor development. With more and more data available on its expression in tumors it is already clear that this lectin is an important target for cancer diagnostics and treatment.Its first involvement in cancer was associated with cell adhesion. Later on the lectin was shown to induce apoptosis of activated T-cells, and experiments have confirmed that cancer cells express galectin-1 also to protect themselves from immune response. Latest finding is a participation of galectin-1 in tumor angiogenesis as an angiogenic factor. In most of the cases described, galectin-1 is up-regulated in tumors in comparison to normal tissue. Moreover, the presence of the lectin is associated with metastases and poor prognosis of disease outcome.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.