Human GAL3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA303HU
MSPP-SEA303HUEA 630 USD
MSPP-SEA303HU
Human GAL3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human GAL3 (Galectin 3). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.054 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Galectin-3 contains a carboxyl-terminal lectin domain and an amino-terminal non-lectin part consisting primarily of short tandem repeats. It is widely distributed in tissues and found in epithelial cells, fibroblasts, dendritic cells, and inflammatory cells. In many cell types studied, galectin-3 is present diffusely in the cytoplasm, but is also localized to the nucleus and subcellular structures, such as mitochondria, phagosomes and exosomes, under specific conditions. It is secreted by various cell types, including monocytes, macrophages, and epithelial cells, and the extracellular protein can bind to a large number of different glycoconjugates on the cell surfaces and extracellular matrices.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.