Human NPTX1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA298HU
MSPP-SEA298HUEA 630 USD
MSPP-SEA298HU
Human NPTX1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human NPTX1 (Neuronal Pentraxin I). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.057 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Neuronal pentraxin 1 is a secreted member of the pentraxin family identified first in rats as a protein that may mediate the uptake of synaptic material and the presynaptic snake venom toxin, taipoxin. Rat, mouse, and human sequences are highly conserved. NPTX1 is expressed almost exclusively in central neurons of the nervous system. NPTX1 is expressed constitutively in the murine CNS and is not modulated by administration of bacterial lipopolysaccharides (as is the expression of some other pentraxin family members. NPTX1 expression is induced in neonatal rat brain following perinatal hypoxia-ischemia, and NPTX1 gene silencing has been shown to be neuroprotective against hypoxia-induced neuronal cell death.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.