Human HSP47 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA266HU
MSPP-SEA266HUEA 630 USD
MSPP-SEA266HU
Human HSP47 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human HSP47 (Heat Shock Protein 47). The assay range is from 125 to 8000 pg/ml (Sandwich kit) with a sensitivity of 46 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Heat shock protein 47 is a member of the serpin superfamily of serine proteinase inhibitors. Its expression is induced by heat shock. The protein localizes to the endoplasmic reticulum lumen and binds collagen; thus it is thought to be a molecular chaperone involved in the maturation of collagen molecules. Autoantibodies to this protein have been found in patients with rheumatoid arthritis. Colligins are characterized by an amino acid structure that includes an N-terminal hydrophobic signal sequence and 2 putative N-linked oligosaccharide attachment sites (Clarke et al. 1991). Colligins also have a C-terminal RDEL sequence that acts as an endoplasmic reticulum (ER) retention sequence. Other features permit the colligin-binding protein of ER to be classified as a serpin (serine-arginine protease inhibitor).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.