Rat b2M ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA260RA
MSPP-SEA260RAEA 684 USD
MSPP-SEA260RA
Rat b2M ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat b2M (Beta to 2 to Microglobulin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.134 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
β2 microglobulin lies lateral to the α3 chain on the cell surface. Unlike α3, β2 has no transmembrane region. Directly above β2 lies the α1 chain, which itself is lateral to the α2. β2 microglobulin associates not only with the alpha chain of MHC class I molecules, but also with class I-like molecules such as CD1 and Qa. An additional function is association with the HFE protein, together regulating the expression of hepcidin in the liver which targets the iron transporter ferroportin on the cytoplasmic membrane of enterocytes and macrophages for degradation resulting in decreased iron uptake from food and iron release from recycled red blood cells respectively. Loss of this function causes iron excess and hemochromatosis.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.