Mouse C1INH ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA235MU
MSPP-SEA235MUEA 648 USD
MSPP-SEA235MU
Mouse C1INH ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse C1INH (Complement 1 Inhibitor). The assay range is from 15.6 to 1000 ng/ml (Sandwich kit) with a sensitivity of 6.5 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
C1-INH is a critically important protein that controls activation of multiple plasma mediator pathways. This protein, a member of the serine protease inhibitor (serpin) group, originally was described as an inhibitor of C1. It binds stoichiometrically to the active sites on both C1r and C1s to form a complex C1-INH-C1r-C1s-C1-INH and thus inhibits activated C1. In addition, C1-INH has been reported to remove the intact C1qrs complex from an activating surface and to inhibit autoactivation of C1. C1-INH is a known inhibitor of kinin generating (kallikrein), fibrinolytic (plasmin), and contact activation (intrinsic) pathway of the coagulation cascade. Recently, it has been shown to be an inhibitor of the mannan-binding lectin pathway of complement activation, inhibiting mannan-binding lectin-associated serine proteases (MASPs) in that pathway.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.