Human PF4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA172HU
MSPP-SEA172HUEA 560 USD
MSPP-SEA172HU
Human PF4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PF4 (Platelet Factor 4). The assay range is from 1.56 to 100 pg/ml (Sandwich kit) with a sensitivity of 0.65 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Platelet factor 4 (PF4) is a small cytokine belonging to the CXC chemokine family. This chemokine is released from alpha-granules of activated platelets during platelet aggregation, and promotes blood coagulation by moderating the effects of heparin-like molecules. Due to these roles, it is predicted to play a role in wound repair and inflammation. It is usually found in a complex with proteoglycan. PF4 is chemotactic for neutrophils, fibroblasts and monocytes, and interacts with a splice variant of the chemokine receptor CXCR3, known as CXCR3B. PF4 is the antigen in heparin-induced thrombocytopenia, an idiosyncratic autoimmune reaction to the administration of the anticoagulant heparin. PF4 autoantibodies have also been found in patients with thrombosis and features resembling HIT but no prior administration of heparin.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.