Human OLFM4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA162HU
MSPP-SEA162HUEA 630 USD
MSPP-SEA162HU
Human OLFM4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human OLFM4 (Olfactomedin 4). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Olfactomedin 4is an antiapoptotic factor, can promote tumor growth. It promotes proliferation of pancreatic cancer cells by favoring the transition from the S to G2/M phase. It Facilitates cell adhesion. OLFM4 expressed during myeloid lineage development.Strongly expressed in the prostate, small intestine and colon and moderately expressed in the bone marrow and stomach. Highly expressed in pancreatic cancer tissues and shows an elevated expression level during the early S phase of the cell cycle. Also expressed at high levels in stomach cancer and colon cancer tissues. OLFM4 was originally cloned from human myeloblasts and found to be selectively expressed in inflammed colonic epithelium. The protein encoded is a member of the olfactomedin-related protein family. The exact function of this gene has not yet been determined.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.