Rat AFP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA153RA
MSPP-SEA153RAEA 684 USD
MSPP-SEA153RA
Rat AFP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat AFP (Alpha to Fetoprotein). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.127 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
AFP) is a glycoprotein with a molecular weight of between 65,000 and 70,000 daltons including 4% of carbohydrate. During fetal development, AFP maintains high levels in the serum and drops to very low levels throughout the remainder of life. AFP is elevated in the malignant diseases of hepatocellular, testicular nonseminomatous origin, and occasionally of other entodermal origin. AFP may be slightly elevated or persisted in the patients with large hepatic metastases or viral hepatitis. AFP measurement is widely accepted as tumor marker and for monitoring the therapeutic effectiveness of hepatocellular cancer and nonseminomatous testicular cancer .
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.