Rat CEA ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA150RA
MSPP-SEA150RAEA 532 USD
MSPP-SEA150RA
Rat CEA ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat CEA (Carcinoembryonic Antigen). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.118 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Carcinoembryonic antigen (CEA) is a cell-surface glycoprotein with a molecular weight of 180 to 200 kDa, that occurs in high levels in colon epithelial cells during embryonic development. Levels of CEA are significantly lower in colon tissue of adults, but can become elevated when inflammation or tumours arise in any endodermal tissue, including the gastrointestinal tract, respiratory tract, pancreas and breast.An overexpression of CEA protein has been detected in a variety of adenocarcinomas, including gastric, pancreatic, small intestine, colon, rectal, ovarian, breast, cervical and non-small-cell lung cancers. CEA is also expressed by epithelial cells in several non-malignant disorders, including diverticulitis, pancreatitis, inflammatory bowel disease, cirrhosis, hepatitis, bronchitis and renal failure and also in heavy smokers.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.