Mouse VEGFC ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA145MU
MSPP-SEA145MUEA 648 USD
MSPP-SEA145MU
Mouse VEGFC ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse VEGFC (Vascular Endothelial Growth Factor C). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
VEGF-C has been shown to exhibit angiogenic and lymphangiogenic actions. The VEGF family of growth factors and receptors is involved in the development and growth of the vascular endothelial system. Two of its family members, VEGF-C and VEGF-D, regulate the lymphatic endothelial cells via their receptor VEGFR, thus acting as mitogens for these cells. VEGF-C expression is associated with hematological malignancies. Like VEGF it acts as survival factor on leukemia. Together with the expression of their receptors, VEGF and VEGF-C result in the generation of autocrine loops that may support cancer cell survival and proliferation. Further VEGF-C expression has been shown in gastrointestinal tract malignancies where it correlates with lymphatic invasion, lymphnode metastasis and reduced survival.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.