Guinea pig TNFa ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA133GU
MSPP-SEA133GUEA 560 USD
MSPP-SEA133GU
Guinea pig TNFa ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Guinea pig TNFa (Tumor Necrosis Factor Alpha). The assay range is from 7.8 to 500 pg/ml (Sandwich kit) with a sensitivity of 2.8 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
TNFa  is synthesized as a 26 kDa, type II transmembrane protein that is 233 amino acids in length. It contains a 30 amino acid (aa) cytoplasmic domain, a 26 aa transmembrane segment, and a 177 aa extracellular region. TNFa is assembled intracellularly to form a transmembrane, non-covalently-linked homotrimeric protein. The 157 aa residue soluble form of TNFa (sTNF-αis released from the C-terminus of the transmembrane protein through the activity of TNFa-converting enzyme (TACE), a membrane -bound disintegrin metalloproteinase. Rat cells known to express TNF-αinclude B cells, colonic columnar epithelial cells, NK and CD3 CD56 hepatic natural T cells, macrophages, monocytes and monocyte-derived dendritic cells, CD4 and CD8 T cells, mast cells, neutrophils, keratinocytes, plasma cells, and adipocytes.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.