Rabbit TGFb1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA124RB
MSPP-SEA124RBEA 684 USD
MSPP-SEA124RB
Rabbit TGFb1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rabbit TGFb1 (Transforming Growth Factor Beta 1). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
TGF-β is capable of producing a variety of effects and virtually all cell types respond to this factor in some way. The inappropriate presence of active TGF-β1 has been implicated in a variety of pathological conditions Because of the necessity for regulating its activity tightly, TGF-β is secreted by cells in the form of an inactive complex. This complex consists of TGF-β1 associated non-covalently with a protein designated the latency associated peptide (LAP). TGF-β1 and LAP represent components of a pro-peptide that is cleaved in a post-golgi compartment prior to secretion. LAP and TGF-β1 each consist of a disulfide-linked homodimer and the association of these two components renders TGF-β1 inactive and inaccessible to anti-TGF-β antibodies.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.