Human SCF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA120HU
MSPP-SEA120HUEA 468 USD
MSPP-SEA120HU
Human SCF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human SCF (Stem Cell Factor). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Stem cell factor (SCF;MGF) is a cytokine which binds CD117 (c-Kit). SCF is also known as 'steel factor' or 'c-kit ligand'. SCF exists in two forms, cell surface bound SCF and soluble (or free) SCF. Soluble SCF is produced by the cleavage of surface bound SCF by metalloproteases.SCF is a growth factor important for the survival, proliferation, and differentiation of hematopoietic stem cells and other hematopoietic progenitor cells. One of its roles is to change the BFU-E(burst-forming unit-erythroid) cells, which are the earliest erythrocyte precursors in the erythrocytic series, into the CFU-E (colony-forming unit-erythroid).  SCF, along with bFGF (basic fibroblast growth factor) and LIF (leukemia inhibitory factor), prevents spontaneous differentiation of primitive embryonic stem cells in cell culture.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.