Human SOD3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA117HU
MSPP-SEA117HUEA 630 USD
MSPP-SEA117HU
Human SOD3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human SOD3 (Superoxide Dismutase 3, Extracellular). The assay range is from 125 to 8000 pg/ml (Sandwich kit) with a sensitivity of 55 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Extracellular superoxide dismutase [Cu-Zn] encoded by the SOD3, isa member of the superoxide dismutase (SOD) protein family. SODs are antioxidant enzymes that catalyze the dismutation of two superoxide radicals into hydrogen peroxide and oxygen. Extracellular Superoxide Dismutase 3,is thought to protect the brain, lungs, and other tissues from oxidative stress. The protein is secreted into the extracellular space and forms a glycosylated homotetramer that is anchored to the extracellular matrix (ECM) and cell surfaces through an interaction with heparan sulfate proteoglycan and collagen. A fraction of the protein is cleaved near the C-terminus before secretion to generate circulating tetramers that do not interact with the ECM.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.