Human CKM ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA109HU
MSPP-SEA109HUEA 630 USD
MSPP-SEA109HU
Human CKM ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CKM (Creatine Kinase, Muscle). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.63 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
CKM is a cytoplasmic enzyme involved in cellular energy homeostasis. The encoded protein reversibly catalyzes the transfer of "energy-rich" phosphate between ATP and creatine and between phospho-creatine and ADP. Its functional entity is a MM-CK homodimer in striated (sarcomeric) skeletal and cardiac muscle.In heart, in addition to the MM-CK homodimer also the heterodimer consisting of one muscle (MM-CK) and one brain-type (MB-CK) subunit is expressed. This latter is an important serum marker for myocardial infarction, for it is released from damaged myocardial cells into the blood where it can be detected by clinical chemistry. Since no other organ than cardiac muscle expresses significant levels of MB-CK, this clinical test is still fairly reliable, together with troponin-T, for diagnosing a myocardial infarction.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.