Human OPG ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA108HU
MSPP-SEA108HUEA 560 USD
MSPP-SEA108HU
Human OPG ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human OPG (Osteoprotegerin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Osteoprotegerin (OPG) is a cytokine, which can inhibit the production of osteoclasts. It is a member of the tumor necrosis factor (TNF) receptor superfamily. It is a basic glycoprotein comprising 401 amino acid residues arranged into seven structural domains. It is found as either a 60 kDa monomer or 120 kDa dimer linked by disulfide bonds. Osteoprotegerin inhibits the differentiation of osteoclast precursors into osteoclasts and also regulates the resorption of osteoclasts in vitro and in vivo. Osteoprotegerin is a RANK homolog, and works by binding to RANKL on osteoblast/stromal cells, thus blocking the RANKL-RANK ligand interaction between osteoblast/stromal cells and osteoclast precursors. This has the effect of inhibiting the differentiation of the osteoclast precursor into a mature osteoclast.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.