Human MMP3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA101HU
MSPP-SEA101HUEA 468 USD
MSPP-SEA101HU
Human MMP3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human MMP3 (Matrix Metalloproteinase 3). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
MMP-3 is secreted from the cells as a proenzyme. The proenzyme has been shown to stimulate plasminogen activation. The N-terminal pro-domain contains the cysteine switch motif conserved in MMPs that maintains MMP-3 in the latent state. Activation of the proenzyme results in the removal of the pro-domain. MMP-3 activation can be achieved in vitro by proteases such as itself, chyrotrypsin, neutrophil elastase and plasma kallikrein, and by mercury compounds. The resulting active enzyme consists of a catalytic domain with a zinc-binding motif conserved in metzincins. A short hinge peptide links the catalytic domain to the C-terminal hemopexin-like domain. The active MMP-3 is capable of cleaving types III, IV, IX and X collagen, aggrecan, fibronectin, laminin, IGFBP-3, serpins, and IL-1.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.