Human MIP3b ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA096HU
MSPP-SEA096HUEA 630 USD
MSPP-SEA096HU
Human MIP3b ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human MIP3b (Macrophage Inflammatory Protein 3 Beta). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Chemokine (C-C motif) ligand 19 (CCL19) is a small cytokine belonging to the CC chemokine family that is also known as EBI1 ligand chemokine (ELC) and macrophage inflammatory protein-3-beta (MIP-3-beta). CCL19 is expressed abundantly in thymus and lymph nodes, with moderate levels in trachea and colon and low levels in stomach, small intestine, lung, kidney and spleen. The gene for CCL19 is located on human chromosome 9. This chemokine elicits its effects on its target cells by binding to the chemokine receptor chemokine receptor CCR7. It attracts certain cells of the immune system, including dendritic cells and antigen-engaged B cells, CCR7 effector-memory T-Cells.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.