Human LIF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA085HU
MSPP-SEA085HUEA 468 USD
MSPP-SEA085HU
Human LIF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human LIF (Leukemia Inhibitory Factor). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.3 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Leukemia inhibitory factor, an interleukin 6 class cytokine, is a protein in cells that affects cell growth and development. LIF derives its name from its ability to induce the terminal differentiation of myeloid leukaemic cells. Other properties attributed to the cytokine include: the growth promotion and cell differentiation of different types of target cells, influence on bone metabolism, cachexia, neural development, embryogenesis and inflammation. LIF is normally expressed in the trophectoderm of the developing embryo, with its receptor LIFR expressed throughout the inner cell mass. As embryonic stem cells are derived from the inner cell mass at the blastocyst stage, removing them from the inner cell mass also removes their source of LIF.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.