Pig LEP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA084PO
MSPP-SEA084POEA 756 USD
MSPP-SEA084PO
Pig LEP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Pig LEP (Leptin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.113 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Leptin is a protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is a 16-kDa protein that plays a critical role in the regulation of body weight by inhibiting food intake and stimulating energy expenditure. Defects in leptin production cause severe hereditary obesity in rodents and humans. In addition to its effects on body weight, leptin has a variety of other functions, including the regulation of hematopoiesis, angiogenesis, wound healing, and the immune and inflammatory response. Leptin acts through the leptin receptor (LEPR), a single-transmembrane-domain receptor of the cytokine receptor family, which is found in many tissues in several alternatively spliced forms. The LEP gene is the human homolog of the gene (ob) mutant in the mouse 'obese' phenotype.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.