Dog IL6 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA079CA
MSPP-SEA079CAEA 720 USD
MSPP-SEA079CA
Dog IL6 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Dog IL6 (Interleukin 6). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
IL-6 is a multifunctional protein produced by lymphoid and non-lymphoid cells, and by normal and transformed cells, including T cells, monocyte/macrophages, fibroblasts, hepatocytes, vascular endothelial cells, cardiac myxomas, bladder cell carcinomas, myelomas, astrogliomas and glioblastomas. The production of IL-6 in these various cells is regulated, either positively or negatively, by a variety of signals including mitogens, antigenic stimulation, lipopolysaccharides, IL-1, TNF, PDGF and viruses. On the basis of its various activities, IL-6 has also been called interferon-β2 (IFN-β2), 26 kDa protein, B-cell stimulatory factor-2 (BSF-2), hybridoma/plasmacytoma growth factor, hepatocyte stimulating factor, cytotoxic T-cell differentiation factor, and macrophage-granulocyte inducing factor 2A (MGI-2A).
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.