Guinea pig IL4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA077GU
MSPP-SEA077GUEA 720 USD
MSPP-SEA077GU
Guinea pig IL4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Guinea pig IL4 (Interleukin 4). The assay range is from 3.12 to 200 pg/ml (Sandwich kit) with a sensitivity of 1.14 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin 4 is a pleiotropic cytokine produced primarily by activated T lymphocytes,mast cells and basophils. IL-4 has multiple immune response-modulating functions on a variety of cell types. It is an important modulator of the differentiation of precursor T helper cells to the Th2 subset that mediates humoral immunity and modulates antibody production. In addition, IL-4 has also been shown to have anti-tumor activity both in vivoand in vitro.The biological effects of IL-4 are mediated by specific cell surface receptor complexes. One type of functional IL-4 receptor complex consists of the IL-4-binding subunit and a second chain, designated the common c chain because it has also been identified as a component of the receptor complexes for IL-4, IL-7, IL-9 and IL-15.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.