Bovine IL4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA077BO
MSPP-SEA077BOEA 756 USD
MSPP-SEA077BO
Bovine IL4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Bovine IL4 (Interleukin 4). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 5.9 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interleukin 4 is a pleiotropic cytokine produced primarily by activated T lymphocytes,mast cells and basophils. IL-4 has multiple immune response-modulating functions on a variety of cell types. It is an important modulator of the differentiation of precursor T helper cells to the Th2 subset that mediates humoral immunity and modulates antibody production. In addition, IL-4 has also been shown to have anti-tumor activity both In vivo and In vitro. The biological effects of IL-4 are mediated by specific cell surface receptor complexes. One type of functional IL-4 receptor complex consists of the IL-4-binding subunit and a second chain, designated the common c chain because it has also been identified as a component of the receptor complexes for IL-4, IL-7, IL-9 and IL-15.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.