Human IL3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA076HU
MSPP-SEA076HUEA 630 USD
MSPP-SEA076HU
Human IL3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL3 (Interleukin 3). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin-3 (IL-3) is an interleukin, a type of biological signal (cytokine) that can improve the body's natural response to disease as part of the immune system. It acts by binding to the Interleukin-3 receptor. IL-3 stimulates the differentiation of multipotent hematopoietic stem cells into myeloid progenitor cells as well as stimulates proliferation of all cells in the myeloid lineage (erythrocytes, thrombocytes, granulocytes, monocytes, and dendritic cells). It is secreted by activated T cells to support growth and differentiation of T cells from the bone marrow in an immune response. Interleukin-3 is a hematopoietic colony-stimulating factor that is capable of supporting the proliferation of a broad range of hematopoietic cell types. Interleukin-3 also has neurotrophic activity.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.