Human IL17 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA063HU
MSPP-SEA063HUEA 630 USD
MSPP-SEA063HU
Human IL17 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL17 (Interleukin 17). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.8 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin 17 (IL-17; also known as IL-17A) is a 30 to 35 kDa variably glycosylated homodimeric protein that belongs to a unique family of cysteine-knot related proteins. Its sequence was originally isolated from an activated rodent hybridoma and termed CTLA-8. It is synthesized as a 155 amino acid (aa) precursor that contains a 23 aa signal sequence and a 15 kDa, 132 aa mature segment. Although there are two intrachain disulfide bonds that create a ring reminiscent of those found in cysteine-knot proteins, the actual closed knot structure does not appear to form. IL-17 has one potential N-linked glycosylation site.IL-17, a T cell-derived proinf lammatory cytokine, is produced by TCRα/β+CD4-CD8- thymocytes, as well as by activated CD4+ and CD4+CD45RO+ memory T cells.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.