Human IL15 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA061HU
MSPP-SEA061HUEA 468 USD
MSPP-SEA061HU
Human IL15 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL15 (Interleukin 15). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.6 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin 15 (IL-15) is a cytokine with structural similarity to IL-2 that is secreted by mononuclear phagocytes (and some other cells) following infection by virus(es). This cytokine induces cell proliferation of natural killer cells; cells of the innate immune system whose principal role is to kill virally infected cells. The protein encoded by this gene is a cytokine that regulates T and natural killer cell activation and proliferation. This cytokine and interleukin 2 share many biological activities. They are found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between this cytokine and IL2.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.