Human IL13 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA060HU
MSPP-SEA060HUEA 630 USD
MSPP-SEA060HU
Human IL13 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL13 (Interleukin 13). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Interleukin 13 (IL-13) is produced primarily by activated T lymphocytes. It shows approximately 30% amino acid (aa) sequence identity with IL-4 and shares many biological properties with IL-4. IL-13 has multiple effects on the differentiation and functions of monocytes/macrophages. It can suppress the cytotoxic functions of monocytes/macrophages and the production of proinflammatory cytokines by monocytes/macrophages. The biological effects of IL-13 are mediated by specific high-affinity cell surface receptor complexes. The functional IL-13 receptor complex has been shown to consist of the low-affinity IL-13 receptor α chain and the IL-4 receptor α subunit (IL-4 Rα). This IL-13 receptor complex may also serve as an alternate high-affinity IL-4 receptor complex in IL-4 responsive cells that lack the γ chain.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.