Bovine IL10 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA056BO
MSPP-SEA056BOEA 756 USD
MSPP-SEA056BO
Bovine IL10 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Bovine IL10 (Interleukin 10). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 6.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interleukin-10 is a small protein known as a cytokine that functions as an important regulator of the immune system.  Although IL-10 is known to have many different roles in the immune system, its two major activities include inhibition of cytokine production by macrophages and inhibition of the accessory functions of macrophages during T cell activation. The effects of these actions cause IL-10 to play mainly an anti-inflammatory role in the immune system. IL-10 was originally known as the cytokine synthesis inhibiting factor (CSIF), and the discovery of this protein was based on its biological activity. IL-10 is mainly produced by the Th2 subset of CD4+ helper cells.  However, it is also produced by some activated B cells, some Th1 cells, activated macrophages, and some nonhematopoietic sources.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.