Human IGFBP2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA053HU
MSPP-SEA053HUEA 498 USD
MSPP-SEA053HU
Human IGFBP2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IGFBP2 (Insulin Like Growth Factor Binding Protein 2). The assay range is from 62.5 to 4000 pg/ml (Sandwich kit) with a sensitivity of 27.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Insulin-like growth factor binding protein-2 (IGFBP-2) is one of six characterized IGFBPs which specifically bind and modulate the actions of IGF-I and IGF-II. The binding affinity of IGFBP-2 for IGF-II is estimated to be significantly higher than for IGF-I, suggesting a role in regulation of IGF-II action. Studies in the fetal sheep indicate that IGFBP-2 is produced in many tissues, apparently coincident with IGF-II expression. Recent studies of serum IGFBP-2 levels have shown that, unlike for IGFBP-1, there is negligible variability during the day and, unlike IGFBP-1 and IGFBP-3, virtually no change during puberty. IGFBP-2 has also been identified in other body fluids such as cerebrospinal, amniotic and coelomic fluids, urine and seminal fluid.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.