Rat IFNg ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA049RA
MSPP-SEA049RAEA 465 USD
MSPP-SEA049RA
Rat IFNg ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat IFNg (Interferon Gamma). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interferon gamma (IFN-gama ) is a multifunctional protein first observed as an antiviral activity in cultures of Sindbis virus-infected leukocytes stimulated by PHA. Produced by T lymphocytes and natural killer (NK) cells, IFN-γ is now known to be both an inhibitor of viral replication and a regulator of numerous immunological functions. IFN-γ influences the class of antibody produced by B cells, upregulates class I and II MHC complex antigens and increases. the efficiency of macrophage-mediated killing of intracellular parasites. Most of the activities attributed to IFN-γ are believed to be mediated by IFN-γ-induced proteins. The appearance of such proteins is a consequence of IFN-γ binding to a specific receptor that is distinct from the receptor for IFN-αandβ.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.