Human IFNg ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA049HU
MSPP-SEA049HUEA 488 USD
MSPP-SEA049HU
Human IFNg ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IFNg (Interferon Gamma). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interferon gamma (IFN-gama ) is a multifunctional protein first observed as an antiviral activity in cultures of Sindbis virus-infected leukocytes stimulated by PHA. Produced by T lymphocytes and natural killer (NK) cells, IFN-γ is now known to be both an inhibitor of viral replication and a regulator of numerous immunological functions. IFN-γ influences the class of antibody produced by B cells, upregulates class I and II MHC complex antigens and increases. the efficiency of macrophage-mediated killing of intracellular parasites. Most of the activities attributed to IFN-γ are believed to be mediated by IFN-γ-induced proteins. The appearance of such proteins is a consequence of IFN-γ binding to a specific receptor that is distinct from the receptor for IFN-αandβ.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.