Human GM-CSF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA045HU
MSPP-SEA045HUEA 498 USD
MSPP-SEA045HU
Human GM-CSF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human GM-CSF (Colony Stimulating Factor 2, Granulocyte Macrophage). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.2 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Granulocyte-macrophage colony stimulating factor (GM-CSF) was initially identified by its ability to stimulate the formation of granulocyte and macrophage colonies from hematopoietic progenitor cells in semi-solid media. In vivoand in vitro studies have now demonstrated that GM-CSF is a pleiotropic cytokine that induces myeloid precursor cells to proliferate and differentiate into neutrophils, monocyte/macrophages and eosinophils. GM-CSF has also been shown to stimulate the proliferation and differentiation of erythroid and megakaryocyte progenitor cells. In addition, GM-CSF is a survival factor for and activates the effector functions of mature neutrophils, monocytes/macrophages and eosinophils.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.