Human GH ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA044HU
MSPP-SEA044HUEA 398 USD
MSPP-SEA044HU
Human GH ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human GH (Growth Hormone). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Somatotropin is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The gene, along with four other related genes, is located at the growth hormone locus on chromosome 17 where they are interspersed in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. The five genes share a remarkably high degree of sequence identity. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. This particular family member is expressed in the pituitary but not in placental tissue as is the case for the other four genes in the growth hormone locus.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.