Human CXCL1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA041HU
MSPP-SEA041HUEA 468 USD
MSPP-SEA041HU
Human CXCL1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CXCL1 (Chemokine (C-X-C Motif) Ligand 1). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
CXCL1 is secreted by human melanoma cells, has mitogenic properties and is implicated in melanoma pathogenesis.CXCL1 is expressed by macrophages, neutrophils and epithelial cells,and has neutrophil chemoattractant activity. CXCL1 plays a role in spinal cord development by inhibiting the migration of oligodendrocyte precursors and is involved in the processes of angiogenesis, inflammation, wound healing, and tumorigenesis. This chemokine elicits its effects by signaling through the chemokine receptor CXCR2.The gene for CXCL1 is located on human chromosome 4 amongst genes for other CXC chemokines.An initial study in mice showed evidence that CXCL1 decreased the severity of multiple sclerosis and may offer a neuro-protective function.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.