Rat CTGF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEA010RA
MSPP-SEA010RAEA 684 USD
MSPP-SEA010RA
Rat CTGF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat CTGF (Connective Tissue Growth Factor). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.1 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
CTGF (connective tissue growth factor) is a cysteine-rich, matrix-associated, heparin-binding protein. in vitro, CTGF mirrors some of the effects of TGF beta on skin fibroblasts, such as stimulation of extracellular matrix production, chemotaxis, proliferation and integrin expression. CTGF can promote endothelial cell growth, migration, adhesion and survival and is thus implicated in endothelial cell function and angiogenesis. CTGF binds to perlecan, a proteoglycan which has been localised in synovium, cartilage and numerous other tissues.CTGF has been implicated in extracellular matrix remodelling in wound healing, scleroderma and other fibrotic processes, as it is capable of upregulating both matrix metalloproteinases and their inhibitors.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.