Human AREG ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SCA006HU
MSPP-SCA006HUEA 840 USD
MSPP-SCA006HU
Human AREG ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human AREG (Amphiregulin). The assay range is from 1.37 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 0.59 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Amphiregulin is a member of the EGF family of proteins. Amphiregulin is a major autocrine growth factor for cultured human keratinocytes and probably plays a role in the aberrant keratinocyte growth of hyperproliferative disorders.The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype.Amphiregulin, also known as AREG.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.