Jumbo Clamp Holder, Cole-Parmer
Supplier: Cole Parmer
08021-62
MSPP-SC0802162EA 76.8 USD
MSPP-SC0802162
Jumbo Clamp Holder, Cole-Parmer
Clamps General Purpose Clamps
Connect rods and/or clamps to a frame, or build a custom lab frame.
  • Use with 0.5" (12.7 mm) diameter frame rods
  • Connectors and holders do not include rods
Ideal for holding clamps or ring stands to lab-frames, at a 90° angle. Clamp accommodates rod diameters up to 0.83".