FlipMate® Filtration Assembly, Environmental Express
Supplier: ENVIRONMENTAL EXPRESS MS
SC0601
MSPP-SC0601PK 209.78 USD
MSPP-SC0601
FlipMate® Filtration Assembly, Environmental Express
Funnels Filtration Funnels Filtration Apparatus Glass
Simple screw on filtration assembly is designed for 50 ml digestion cup filtration.
  • Made of Poly ether sulfone (PES)
  • Available with prefilter