Support Stand, Cole-Parmer
Supplier: Cole Parmer
04712-92
MSPP-SC0471292EA 133.88 USD
MSPP-SC0471292
Support Stand, Cole-Parmer
Support Stands
Enhance the performance of your ultrasonic processor.
  • Compatible with Cole-Parmer 500 and 700 W ultrasonic processors
  • Durable cast-iron base and stainless steel rod for harsh applications
Base measures 9×5-1/2" (229 x 140 mm). Rod measures 24×1/2" (609×13 mm).