Tailveiner® Rat Restrainers, Braintree Scientific
Supplier: 0000018294
RTV-190 RTV-RED-180
MSPP-RTV190EA 390.77 USD
MSPP-RTV190 MSPP-RTV-R180
Tailveiner® Rat Restrainers, Braintree Scientific
Animal Supplies Animal Restraints
The Tailveiner® is the first easy-to-use mouse and rat restrainer specifically designed for tail vein injections.
  • Slotted tube permits the rat or mouse to be quickly pulled in by the tail
  • Suitable for intravenous injections in small animals
The animal is restrained by a plug which slides easily to accommodate any size rat or mouse, but locks automatically when handle is released. After dosing, the handle is lifted and the plug is easily removed.