Human Recombinant MUC13 (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPP861HU01
MSPP-RPP861HU1EA 436 USD
MSPP-RPP861HU1
Human Recombinant MUC13 (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a MUC13 recombinant protein (prokaryotic), Human is sequencing from Ser174~Gln404 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
The rodent and human MUC13 cDNAs contain a series of CA dinucleotide repeats and an alternative polyadenylation signal. The deduced 512-amino acid human protein has a calculated molecular mass of 54.7 kDa. Both human and rodent MUC13 have an N-terminal signal peptide, followed by a serine/threonine-rich mucin repeat domain, a cysteine-rich EGF-like domain, a SEA module, 2 more cysteine-rich EGF-like domains, a transmembrane region, and a cytoplasmic tail. The mucin repeat domain is predicted to be heavily O-glycosylated, and MUC13 also has sites for N-glycosylation and phosphorylation. Northern blot analysis detected highest expression of a 3.2-kb transcript in colon and trachea, with moderate levels in kidney, small intestine, appendix, and stomach, and low levels in several other tissues.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.