Mouse Recombinant WNT10A (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPP358MU01
MSPP-RPP358MU1EA 460 USD
MSPP-RPP358MU1
Mouse Recombinant WNT10A (prokaryotic)(from E. coli)
Proteins and Peptides
This is a WNT10A recombinant protein (prokaryotic), Mouse is sequencing from Ser108~Pro343 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
WNT10A is a member of the WNT gene family. It is strongly expressed in the cell lines of promyelocytic leukemia and Burkitt's lymphoma. In addition, it and another family member, the WNT6 gene, are strongly coexpressed in colorectal cancer cell lines. The gene overexpression may play key roles in carcinogenesis through activation of the WNT-beta-catenin-TCF signaling pathway. This gene and the WNT6 gene are clustered in the chromosome 2q35 region. WNT10A encodes a deduced 417-amino acid peptide containing residues conserved among WNTs and 2 N-linked glycosylation sites. WNT10A shares 59.2% amino acid sequence identity with human WNT10B.like WNT1 and WNT10B, WNT10A and WNT6 may play key roles in human carcinogenesis.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.