Human Recombinant TAS1R3 (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPF793HU01
MSPP-RPF793HU1EA 450 USD
MSPP-RPF793HU1
Human Recombinant TAS1R3 (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a TAS1R3 recombinant protein (prokaryotic), Human is sequencing from Ala21~Leu570 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
The TAS1R3 gene encodes the human homolog of mouse Sac, a major determinant of differences between sweet-sensitive and -insensitive mouse strains in their responsiveness to sucrose, saccharine, and other sweeteners. The ability to taste the sweetness of carbohydrate-rich foodstuffs has a critical role in the nutritional status of humans. Although several components of bitter transduction pathways had been identified, the receptors and other sweet transduction elements remained unknown at the time of studies by Max et al. (2001). The Sac locus in mouse, mapped to the distal end of chromosome 4, is the major determinant of differences between sweet-sensitive and -insensitive strains of mice in their responsiveness to saccharine, sucrose, and other sweeteners .
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.