Human Recombinant PGLYRP1 (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPD937HU01
MSPP-RPD937HU1EA 410 USD
MSPP-RPD937HU1
Human Recombinant PGLYRP1 (prokaryotic) (from E.coli)
Proteins and Peptides
This is a PGLYRP1 recombinant protein (prokaryotic), Human is sequencing from Gln22~Pro196 with 97 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Innate nonself immune recognition relies on structures common among invading microbes, a process termed pattern recognition. Peptidoglycan is a fundamental component of the bacterial cell wall and is thus a candidate for a pattern recognized by the immune system.
The deduced human PGLYRP protein has 196 amino acids. Both the human and mouse PGLYRP proteins share 43% sequence identity with the T. ni PGLYRP protein. Recombinant mouse Pglyrp protein expressed in insect cells possessed an affinity for peptidoglycan. Dot blot analysis of a number of human tissue mRNAs detected strong expression in bone marrow and weak expression in lung, kidney, liver, small intestine, spleen, thymus, peripheral leukocyte, and fetal spleen.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.