Human Recombinant CHRNa3 (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPD143HU01
MSPP-RPD143HU1EA 300 USD
MSPP-RPD143HU1
Human Recombinant CHRNa3 (prokaryotic)(from E. coli)
Proteins and Peptides
This is a CHRNa3 recombinant protein (prokaryotic), Human is sequencing from Ser32~Leu240 with 95 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Neuronal acetylcholine receptor subunit alpha-3 is a member of the nicotinic acetylcholine receptor family of proteins. Members of this family of proteins form pentameric complexes comprised of both alpha and beta subunits. This locus encodes an alpha-type subunit, as it contains characteristic adjacent cysteine residues. The encoded protein is a ligand-gated ion channel that likely plays a role in neurotransmission. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer. Alternatively spliced transcript variants have been described.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.