Human Recombinant HBa1 (Prokaryotic) (from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPD090HU01
MSPP-RPD090HU1EA 410 USD
MSPP-RPD090HU1
Human Recombinant HBa1 (Prokaryotic) (from E. coli)
Proteins and Peptides
This is a HBa1 recombinant protein (prokaryotic), Human is sequencing from Met1~Arg142 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'-zeta-pseudozeta-mu-pseudoalpha-1-alpha-2-alpha-1-theta-3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.