Human Recombinant TMPRSS2 (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPC795HU01
MSPP-RPC795HU1EA 410 USD
MSPP-RPC795HU1
Human Recombinant TMPRSS2 (prokaryotic)(from E. coli)
Proteins and Peptides
This is a TMPRSS2 recombinant protein (prokaryotic), Human is sequencing from Ser284~Gly492 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Transmembrane protease, serine 2 protein that belongs to the serine protease family. The encoded protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. This gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.