Human Recombinant BANF1 (prokaryotic) (from E.coli)
Supplier: CLOUD-CLONE CORP MS
RPC336HU01
MSPP-RPC336HU1EA 385 USD
MSPP-RPC336HU1
Human Recombinant BANF1 (prokaryotic) (from E.coli)
Proteins and Peptides
This is a BANF1 recombinant protein (prokaryotic), Human is sequencing from Thr2~Leu89 with 95 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Barrier-to-autointegration factor was identified by its ability to protect retroviruses from intramolecular integration and therefore promote intermolecular integration into the host cell genome. The endogenous function of the protein is unknown.
The protein forms a homodimer which localizes to the nucleus and is specifically associated with chromosomes during mitosis. This protein binds to DNA in a non-specific manner and studies in rodents suggest that it also binds to lamina-associated polypeptide 2, a component of the nuclear lamina. It also associates with the LEM Domain containing proteins LAP2, Emerin, and MAN1.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.