Human Recombinant TERT (prokaryotic)(from E. coli)
Supplier: CLOUD-CLONE CORP MS
RPC241HU01
MSPP-RPC241HU1EA 410 USD
MSPP-RPC241HU1
Human Recombinant TERT (prokaryotic)(from E. coli)
Proteins and Peptides
This is a TERT recombinant protein (prokaryotic), Human is sequencing from Arg787~Arg1084 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee
Telomerase reverse transcriptase is a catalytic subunit of the enzyme telomerase. Its absence (usually as a result of a chromosomal mutation) is associated with the disorder Cri du chat.Telomerase is a ribonucleoprotein polymerase that maintains telomere ends by addition of the telomere repeat TTAGGG. The enzyme consists of a protein component with reverse transcriptase activity, encoded by this gene, and an RNA component which serves as a template for the telomere repeat. Telomerase expression plays a role in cellular senescence, as it is normally repressed in postnatal somatic cells resulting in progressive shortening of telomeres. Deregulation of telomerase expression in somatic cells may be involved in oncogenesis.
Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.